missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Monobasic, ≥98%, MP Biomedicals

Supplier:  MP Biomedicals Inc 0219485091

 View more versions of this product

Catalog No. ICN19485091


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Phosphate Monobasic
7558-80-7
≥98%
MFCD00003527,MFCD00146206
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
sodium dihydrogen phosphate
23672064
120
Anhydrous
1 kg
H2NaO4P
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate
[Na+].OP(O)([O-])=O
119.98
CHEBI:37585

Chemical Identifiers

7558-80-7
119.98
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
23672064
sodium dihydrogen phosphate
H2NaO4P
MFCD00003527,MFCD00146206
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate
CHEBI:37585
[Na+].OP(O)([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions