missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic, Puriss., meets analytical spec. of BP, Ph. Eur., USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance), Honeywell Fluka™
Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Supplier: Honeywell-Fluka 042764X5KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Sodium Phosphate Dibasic | |
| 7558-79-4 | |
| HNa2O4P | |
| MFCD00003496 | |
| Very Soluble in hot water, 8 parts soluble in water , insoluble in alcohol | |
| [Na+].[Na+].OP([O-])([O-])=O | |
| 141.96 | |
| CHEBI:34683 | |
| 98 to 100.5% (calc. to the dried substance) |
| Meets analytical specification of Ph.Eur.BP, USP, FCC | |
| 4 x 5 kg | |
| Na2HPO4 | |
| disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| disodium;hydrogen phosphate | |
| 24203 | |
| 141.96 | |
| puriss. |
Chemical Identifiers
| 7558-79-4 | |
| 141.96 | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| 24203 | |
| disodium;hydrogen phosphate |
| HNa2O4P | |
| MFCD00003496 | |
| disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
| CHEBI:34683 | |
| [Na+].[Na+].OP([O-])([O-])=O |
Safety and Handling
EINECSNumber : 231-448-7
RTECSNumber : WC4500000