missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic, Puriss., meets analytical spec. of BP, Ph. Eur., USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance), Honeywell Fluka™

Catalog No. p-7071721
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
4 x 5 kg
5 kg
6 x 1 kg
Catalog No. Quantity
6001841 1 kg
6001843 5 kg
60044552 6 x 1 kg
60044551 4 x 5 kg
4 options

Catalog No. 6001841

Supplier: Honeywell Chemicals 042761KG

Only null left

Puriss., meets analytical specification of BP, Ph. Eur., USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium;hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Anhydrous
Quantity 1 kg
Linear Formula Na2HPO4
Solubility Information Very Soluble in hot water, 8 parts soluble in water , insoluble in alcohol
Formula Weight 141.96g/mol
Percent Purity 98 to 100.5% (calc. to the dried substance)
Grade puriss.
Chemical Name or Material Sodium Phosphate Dibasic, Meets analytical specification of Ph.Eur.BP, USP, FCC
EINECSNumber 231-448-7
RTECSNumber WC4500000