missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate dibasic, ACS Reagent, Honeywell Fluka™

ACS Reagent,≥99.0%

$195.18 - $269.47

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6003204
View Documents
Honeywell-Fluka
S9763100G
100 g
Each for $195.18
Only null left
 
6003209
View Documents
Honeywell-Fluka
S9763500G
500 g
Each for $269.47
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7558-79-4
141.96
BNIILDVGGAEEIG-UHFFFAOYSA-L
24203
[Na+].[Na+].OP([O-])([O-])=O
HNa2O4P
MFCD00003496
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
CHEBI:34683

Specifications

7558-79-4
HNa2O4P
MFCD00003496
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
[Na+].[Na+].OP([O-])([O-])=O
141.96
CHEBI:34683
≥99%
Sodium phosphate dibasic
100 g
Na2HPO4
NONH for all modes of transport
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium;hydrogen phosphate
24203
141.96g/mol
ACS Reagent
Videos
Safety and Handling

Safety and Handling

RTECSNumber : WC4500000

SDS
missing translation for 'documents'

missing translation for 'documents'