missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic, ACS Reagent, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 500 g |
| Linear Formula | Na2HPO4 |
| UN Number | NONH for all modes of transport |
| IUPAC Name | disodium;hydrogen phosphate |
| Formula Weight | 141.96g/mol |
| Percent Purity | ≥99% |
| Grade | ACS Reagent |
| Chemical Name or Material | Sodium phosphate dibasic |
Safety and Handling
| RTECSNumber | WC4500000 |