missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate, dibasic, 98+%, extra pure, anhydrous
CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol
Supplier: Thermo Scientific Chemicals 448140050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sodium phosphate, dibasic | |
| 5 kg | |
| HNa2O4P | |
| MFCD00003496 | |
| disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| disodium hydrogen phosphate | |
| 24203 | |
| 141.96 | |
| Extra Pure |
| 7558-79-4 | |
| 98% | |
| Na2HPO4 | |
| 15,8789 | |
| Solubility in water: 75g/L (20°C). Other solubilities: practically insoluble in alcohol | |
| [Na+].[Na+].OP([O-])([O-])=O | |
| 141.96 | |
| CHEBI:34683 | |
| ≥98% |
Chemical Identifiers
| 7558-79-4 | |
| 141.96 | |
| BNIILDVGGAEEIG-UHFFFAOYSA-L | |
| 24203 | |
| [Na+].[Na+].OP([O-])([O-])=O |
| HNa2O4P | |
| MFCD00003496 | |
| disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
| CHEBI:34683 |
Safety and Handling
EINECSNumber : 231-448-7
RUO – Research Use Only