missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, dibasic, 98+%, extra pure, anhydrous

Catalog No. p-4813654
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC448140010 1 kg
AC448140050 5 kg
AC448140250 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC448140010 Supplier Thermo Scientific Chemicals Supplier No. 448140010
Only null left

CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Quantity 1 kg
Assay Percent Range 98+%
Linear Formula Na2HPO4
Merck Index 15,8789
Solubility Information Solubility in water: 75g/L (20°C). Other solubilities: practically insoluble in alcohol
IUPAC Name disodium;hydrogen phosphate
Formula Weight 141.96
Percent Purity ≥98%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Sodium phosphate, dibasic
Show More Show Less
EINECSNumber 231-448-7

RUO – Research Use Only