missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Persulfate, 1.50 M, Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 72.0, 25.8 |
| CAS Max % | 75.0, 26.86 |
| Grade | Laboratory |
| Packaging | Cubitainer |
| pH | 6 |
| Quantity | 20 L |
| Physical Form | Liquid |
| Concentration or Composition Notes | 1.50Molar |
| Chemical Name or Material | Sodium Persulfate |