missing translation for 'onlineSavingsMsg'
Learn More

Sodium Peroxide, Reagent, ACS, 93%, Spectrum™ Chemical

S1390, 1313-60-6, Na2O2

Supplier:  Spectrum Chemical Mfg Cor S139025KG

 View more versions of this product

Catalog No. 18606696


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Sodium Peroxide, Reagent, ACS is a solid that comes in the form yellow micro beads. It is an inorganic compound that was historically used to bleach wood pulp in order to prepare it for the use in paper manufacture. Today it is most commonly used for certain specialized lab operations such as mineral extraction form ore. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Specifications

Specifications

1313-60-6
2.5 kg
PFUVRDFDKPNGAV-UHFFFAOYSA-N
disodium dioxidanediide
93%
Steel Can
1
Na2O2
[Na+].[Na+].[O-][O-]
77.98
ACS

Chemical Identifiers

1313-60-6
77.98
disodium dioxidanediide
Na2O2
PFUVRDFDKPNGAV-UHFFFAOYSA-N
[Na+].[Na+].[O-][O-]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions