missing translation for 'onlineSavingsMsg'
Learn More

Sodium Peroxide, Reagent, ACS, 93%, Spectrum™ Chemical

Catalog No. p-7158251
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606693 25 g Steel Can
18606694 125 g Steel Can
18606695 500 g Steel Can
18606696 2.5 kg Steel Can
18606697 12 kg Poly Pail
18606698 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18606693 Supplier Spectrum Chemical Mfg Cor Supplier No. S139025GM
Only null left

S1390, 1313-60-6, Na2O2

Spectrum™ Chemical Sodium Peroxide, Reagent, ACS is a solid that comes in the form yellow micro beads. It is an inorganic compound that was historically used to bleach wood pulp in order to prepare it for the use in paper manufacture. Today it is most commonly used for certain specialized lab operations such as mineral extraction form ore. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 1313-60-6
Molecular Formula Na2O2
Molecular Weight (g/mol) 77.98
InChI Key PFUVRDFDKPNGAV-UHFFFAOYSA-N
IUPAC Name disodium dioxidanediide
SMILES [Na+].[Na+].[O-][O-]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 93%
Grade ACS
Packaging Steel Can