accessibility menu, dialog, popup

UserName

NaNaOO

CAS RN 1313-60-6

IUPAC Name: disodium dioxidanediide
Synonyms: sodium peroxide
Molecular Weight (g/mol): 77.98
Molecular Formula: Na2O2
InChi Key: PFUVRDFDKPNGAV-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[O-][O-]
Grade 
  • (3)
  • (3)
  • (6)
  • (4)
  • (1)
  • (1)
  • (4)
  • (6)
Molecular Weight (g/mol) 
  • (6)
  • (12)
  • (10)
Percent Purity 
  • (3)
  • (4)
  • (4)
  • (4)
  • (6)
  • (3)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (8)
  • (1)
Physical Form 
  • (3)
  • (10)
  • (7)
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (10)
  • (1)
  • (4)
  • (1)
  • (10)
  • (12)
  • (3)
  • (3)
  • (6)
  • (4)
  • (1)
  • (1)
  • (4)
  • (6)
  • (6)
  • (12)
  • (10)
  • (3)
  • (4)
  • (4)
  • (4)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (8)
  • (1)
  • (3)
  • (10)
  • (7)
  • (2)
  • (3)
  • (1)