missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium perchlorate, Honeywell Fluka™
ACS Reagent,≥98.0%
$556.34 - $556.34
Chemical Identifiers
| CAS | 7601-89-0 |
|---|---|
| Molecular Formula | ClNaO4 |
| Molecular Weight (g/mol) | 122.436 |
| MDL Number | MFCD00003480 |
| InChI Key | BAZAXWOYCMUHIX-UHFFFAOYSA-M |
| Synonym | sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian |
| PubChem CID | 522606 |
| IUPAC Name | sodium;perchlorate |
| SMILES | [O-]Cl(=O)(=O)=O.[Na+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7601-89-0 | |
| 122.436 | |
| BAZAXWOYCMUHIX-UHFFFAOYSA-M | |
| 522606 | |
| [O-]Cl(=O)(=O)=O.[Na+] |
| ClNaO4 | |
| MFCD00003480 | |
| sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian | |
| sodium;perchlorate |
Specifications
| 7601-89-0 | |
| ClNaO4 | |
| MFCD00003480 | |
| sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian | |
| [O-]Cl(=O)(=O)=O.[Na+] | |
| 122.436 | |
| 122.44g/mol | |
| Sodium perchlorate |
| 500 g | |
| NaClO4 | |
| UN1502 | |
| BAZAXWOYCMUHIX-UHFFFAOYSA-M | |
| sodium;perchlorate | |
| 522606 | |
| ≥98% |
Safety and Handling
P220-P305 + P351 + P338
RTECSNumber : SC9800000