Learn More
Sodium perborate monohydrate, 95%
CAS: 10332-33-9 | BNaO3·H2O | 99.81 g/mol
$59.21 - $674.14
Chemical Identifiers
| CAS | 10332-33-9 |
|---|---|
| Molecular Formula | BNaO3·H2O |
| Molecular Weight (g/mol) | 99.81 |
| MDL Number | MFCD00149230 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC386410250
|
Thermo Scientific Chemicals
386410250 |
25 g | Plastic bottle |
Each for $59.21
|
|
||||
|
AC386412500
|
Thermo Scientific Chemicals
386412500 |
250 g | Plastic bottle |
Each for $240.51
|
|
||||
|
AC386410010
|
Thermo Scientific Chemicals
386410010 |
1 kg | Plastic bottle |
Each for $674.14
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
| 10332-33-9 | |
| BNaO3·H2O | |
| 99.81 | |
| MFCD00149230 |
Specifications
| White | |
| Crystalline Powder | |
| 94% min. (active oxygen) (Iodometry) | |
| MFCD00149230 | |
| 99.81 | |
| 95% | |
| Sodium perborate monohydrate |
| 10332-33-9 | |
| 25 g | |
| BNaO3·H2O | |
| Solubility in water: 15g/L (20°C) | |
| 99.81 | |
| Plastic bottle |
Safety and Handling
GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
May cause respiratory irritation.
Causes serious eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Obtain special instructions before use.
Avoid
GHS Signal Word: Danger
RUO – Research Use Only