missing translation for 'onlineSavingsMsg'
Learn More

Sodium perborate monohydrate, 95%

CAS: 10332-33-9 | BNaO3·H2O | 99.81 g/mol

Supplier:  Thermo Scientific Chemicals 386410250

 View more versions of this product

Catalog No. AC386410250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

Sodium perborate monohydrate
10332-33-9
25 g
BNaO3·H2O
Solubility in water: 15g/L (20°C)
99.81
Plastic bottle
White
Crystalline Powder
94% min. (active oxygen) (Iodometry)
MFCD00149230
99.81
95%

Chemical Identifiers

10332-33-9
BNaO3·H2O
99.81
MFCD00149230
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May damage the unborn child.
Suspected of damaging fertility.
May cause respiratory irritation.
Causes serious eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Obtain special instructions before use.
Avoid

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only