missing translation for 'onlineSavingsMsg'
Learn More

Sodium p-Toluenesulfonate 90.0+%, TCI America™

Catalog No. p-7136180
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T027125G 25 g
T0271500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T027125G Supplier TCI America Supplier No. T027125G
Only null left

Chemical Identifiers

CAS 657-84-1
Molecular Formula C7H7NaO3S
Molecular Weight (g/mol) 194.18
MDL Number MFCD00798566,MFCD00064388
InChI Key KVCGISUBCHHTDD-UHFFFAOYSA-M
Synonym sodium p-toluenesulfonate, sodium 4-methylbenzenesulfonate, sodium tosylate, p-toluenesulfonic acid sodium salt, sodium toluenesulfonate, sodium toluenesulphonate, tosylate, sodium, naxonate hydrotrope, cyclophil sts 70, eltesol st 34
PubChem CID 3720192
IUPAC Name sodium 4-methylbenzene-1-sulfonate
SMILES [Na+].CC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 194.18
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium p-Toluenesulfonate
EINECSNumber (3)-1883&(3)-1900
RTECSNumber XT7350000
TSCA Yes