missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate, For Nitrogen (Nitrite), Certified, 0.0250M ±0.0005M (0.05N), LabChem™

Catalog No. LC247201
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC247201 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC247201 Supplier LabChem Supplier No. LC247201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62-76-0 , 7732-18-5
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Color Colorless
CAS Max % 99.66, 0.34
Linear Formula Na2C2O4
Concentration 0.0250M ±0.0005M (0.05N)
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 134
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Decomposition Information Carbon monoxide; Carbon dioxide
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Sodium Oxalate, For Nitrogen (Nitrite)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature