missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Oxalate, 0.100 M (M/10), 0.200 N (N/5), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 97.0, 1.3 |
| CAS Max % | 100.0, 1.35 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | Neutral |
| Quantity | 120 mL |
| Physical Form | Liquid |
| Concentration or Composition Notes | 0.100Molar,0.200Normal |
| Chemical Name or Material | Sodium Oxalate |