missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate, 0.100 M (M/10), 0.200 N (N/5), Ricca Chemical

Catalog No. 75004
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
75004 120 mL Natural Poly Bottle
750016 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 75004 Supplier Ricca Chemical Company Supplier No. 75004

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62-76-0 , 7732-18-5
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Color Colorless
CAS Min % 97.0, 1.3
CAS Max % 100.0, 1.35
Grade Laboratory
Packaging Natural Poly Bottle
pH Neutral
Quantity 120 mL
Physical Form Liquid
Concentration or Composition Notes 0.100Molar,0.200Normal
Chemical Name or Material Sodium Oxalate