missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Oleate 97.0+%, TCI America™
Supplier: TCI America O0057100G
Specifications
| Sodium Oleate | |
| 235°C | |
| C18H33NaO2 | |
| 100 g | |
| BCKXLBQYZLBQEK-KVVVOXFISA-M | |
| sodium;(Z)-octadec-9-enoate | |
| 23665730 | |
| 304.45 | |
| Crystalline Powder |
| 143-19-1 | |
| White-Yellow | |
| MFCD00004438 | |
| sodium oleic acid | |
| CCCCCCCCC=CCCCCCCCC(=O)[O-].[Na+] | |
| 304.45 | |
| CHEBI:81860 | |
| ≥97.0% (T) |
Chemical Identifiers
| 143-19-1 | |
| 304.45 | |
| BCKXLBQYZLBQEK-KVVVOXFISA-M | |
| 23665730 | |
| sodium;(Z)-octadec-9-enoate |
| C18H33NaO2 | |
| MFCD00004438 | |
| sodium oleic acid | |
| CHEBI:81860 | |
| CCCCCCCCC=CCCCCCCCC(=O)[O-].[Na+] |
Safety and Handling
EINECSNumber : (2)-0611&(7)-0973
RTECSNumber : RK1200000
TSCA : Yes