accessibility menu, dialog, popup

UserName

NaH3COO(Z)

CAS RN 143-19-1

IUPAC Name: sodium (9Z)-octadec-9-enoate
Synonyms: sodium oleate
Molecular Weight (g/mol): 304.45
Molecular Formula: C18H33NaO2
InChi Key: BCKXLBQYZLBQEK-KVVVOXFISA-M
SMILES: [Na+].CCCCCCCC\C=C/CCCCCCCC([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)