missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oleate 97.0+%, TCI America™

Catalog No. p-7134161
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O005725G 25 g
O0057100G 100 g
O0057500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O005725G Supplier TCI America Supplier No. O005725G
Only null left

Chemical Identifiers

CAS 143-19-1
Molecular Formula C18H33NaO2
Molecular Weight (g/mol) 304.45
MDL Number MFCD00004438
InChI Key BCKXLBQYZLBQEK-KVVVOXFISA-M
Synonym sodium oleic acid
PubChem CID 23665730
ChEBI CHEBI:81860
IUPAC Name sodium;(Z)-octadec-9-enoate
SMILES CCCCCCCCC=CCCCCCCCC(=O)[O-].[Na+]

Specifications

Melting Point 235°C
Color White-Yellow
Quantity 25 g
Formula Weight 304.45
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Oleate
EINECSNumber (2)-0611&(7)-0973
RTECSNumber RK1200000
TSCA Yes