missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, 5% (w/w), Ricca Chemical

Catalog No. p-3252391
Change view
Click to view available options
Quantity:
120 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
74974 120 mL Amber Glass Bottle
749732 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74974 Supplier Ricca Chemical Company Supplier No. 74974
Only null left

Chemical Identifiers

CAS 7632-00-0 , 7732-18-5
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

Color Colorless
CAS Min % 93.0, 4.81
CAS Max % 97.0, 5.0
Percent Purity 5%
Grade Laboratory
Packaging Amber Glass Bottle
Solvent or Matrix Aqueous Solution
pH ∼9
Quantity 120 mL
Physical Form Liquid
Concentration or Composition Notes 5% (w/w)
Chemical Name or Material Sodium Nitrite
Show More Show Less