missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Sodium n-Dodecyl Sulfate, 20% Solution (w/v), Calbiochem™,

Catalog No. 428018200ML
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
428018200ML 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 428018200ML Supplier MilliporeSigma™ Supplier No. 428018200ML

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym 20% SDS
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color Colorless
Quantity 200 mL
Packaging Glass Bottle
RTECSNumber WT1050000