missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Methoxide (ca. 5mol/L in Methanol), TCI America™
Supplier: TCI America S0486500ML
Specifications
| Sodium Methoxide (ca. 5mol/L in Methanol) | |
| 124-41-4 | |
| 500 mL | |
| MFCD00012179 | |
| sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 | |
| C[O-].[Na+] | |
| 54.024 | |
| 54.02 |
| Colorless | |
| Liquid | |
| CH3NaO | |
| 1289 | |
| WQDUMFSSJAZKTM-UHFFFAOYSA-N | |
| sodium;methanolate | |
| 10942334 |
Chemical Identifiers
| 124-41-4 | |
| 54.024 | |
| WQDUMFSSJAZKTM-UHFFFAOYSA-N | |
| 10942334 | |
| C[O-].[Na+] |
| CH3NaO | |
| MFCD00012179 | |
| sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 | |
| sodium;methanolate |
Safety and Handling
EINECSNumber : (2)-0203
RTECSNumber : PC3570000
TSCA : Yes