missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Methoxide (ca. 5mol/L in Methanol), TCI America™
$39.14 - $52.19
Chemical Identifiers
| CAS | 124-41-4 |
|---|---|
| Molecular Formula | CH3NaO |
| Molecular Weight (g/mol) | 54.024 |
| MDL Number | MFCD00012179 |
| InChI Key | WQDUMFSSJAZKTM-UHFFFAOYSA-N |
| Synonym | sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 |
| PubChem CID | 10942334 |
| IUPAC Name | sodium;methanolate |
| SMILES | C[O-].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
S0486100ML
|
TCI America
S0486100ML |
100 mL |
Each for $39.14
|
|
|||||
|
S0486500ML
|
TCI America
S0486500ML |
500 mL |
Each for $52.19
|
|
|||||
Chemical Identifiers
| 124-41-4 | |
| 54.024 | |
| WQDUMFSSJAZKTM-UHFFFAOYSA-N | |
| 10942334 | |
| C[O-].[Na+] |
| CH3NaO | |
| MFCD00012179 | |
| sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 | |
| sodium;methanolate |
Specifications
| Colorless | |
| Liquid | |
| CH3NaO | |
| 1289 | |
| WQDUMFSSJAZKTM-UHFFFAOYSA-N | |
| sodium;methanolate | |
| 10942334 | |
| Sodium Methoxide (ca. 5mol/L in Methanol) |
| 124-41-4 | |
| 100 mL | |
| MFCD00012179 | |
| sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755 | |
| C[O-].[Na+] | |
| 54.024 | |
| 54.02 |
Safety and Handling
EINECSNumber : (2)-0203
RTECSNumber : PC3570000
TSCA : Yes