Learn More
Sodium Metaperiodate, ACS Reagent (Sodium Periodate), Reagents
Sodium Metaperiodate, ACS Reagent (Sodium Periodate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C1322700500C2
Specifications
| Sodium Metaperiodate, ACS Reagent (Sodium Periodate) | |
| 7790-28-5 | |
| 500 g | |
| Sodium Tetraoxoiodate(VII) | |
| [Na+].[O-][I](=O)(=O)=O | |
| 213.89 | |
| Glass Bottle |
| White | |
| Solid | |
| INaO4 | |
| JQWHASGSAFIOCM-UHFFFAOYSA-M | |
| sodium periodate | |
| ACS | |
| 3.865 |
Chemical Identifiers
| 7790-28-5 | |
| 213.89 | |
| Sodium Tetraoxoiodate(VII) | |
| [Na+].[O-][I](=O)(=O)=O |
| INaO4 | |
| JQWHASGSAFIOCM-UHFFFAOYSA-M | |
| sodium periodate |