accessibility menu, dialog, popup

UserName

NaOIOOO

CAS RN 7790-28-5

IUPAC Name: sodium periodate
Synonyms: sodium periodate
Molecular Weight (g/mol): 213.89
Molecular Formula: INaO4
InChi Key: JQWHASGSAFIOCM-UHFFFAOYSA-M
SMILES: [Na+].[O-][I](=O)(=O)=O
Grade 
  • (7)
  • (6)
  • (3)
  • (2)
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (7)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (7)
  • (1)
Percent Purity 
  • (2)
  • (13)
  • (4)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (4)
  • (2)
  • (2)
  • (7)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (13)
  • (7)
  • (6)
  • (3)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (7)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (7)
  • (1)
  • (2)
  • (13)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (7)
  • (4)