missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metaperiodate, ACS Reagent (Sodium Periodate), Reagents

Catalog No. p-200020938
Change view
Click to view available options
Quantity:
250 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337704 250 g
01337703 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337704 Supplier Labchem Inc Supplier No. C1322700250C2
Only null left

Sodium Metaperiodate, ACS Reagent (Sodium Periodate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym Sodium Tetraoxoiodate(VII)
IUPAC Name sodium periodate
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Color White
Physical Form Solid
Quantity 250 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 3.865
Chemical Name or Material Sodium Metaperiodate, ACS Reagent (Sodium Periodate)