missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium metaperiodate, 99.0%, for analysis EMSURE™ ACS, Reag. Ph Eur, MilliporeSigma™
Supplier: Technidata 1065971000
Specifications
| 270°CÂ (decomposes) | |
| 5.2 (50Â g/L, H2O, 20°C) | |
| Assay (iodometric): ≥ 99.0%; Assay (out of dried substance): 99.8 to 100.3% | |
| Sodium periodate | |
| JQWHASGSAFIOCM-UHFFFAOYSA-M | |
| sodium periodate | |
| Plastic Bottle |
| 7790-28-5 | |
| 1 kg | |
| INaO4 | |
| 91Â g/L | |
| [Na+].[O-][I](=O)(=O)=O | |
| 213.89 | |
| 3.87Â g/cm3 (at 20°C); Bulk Density: 2900Â kg/m3 |
Chemical Identifiers
| 7790-28-5 | |
| 213.89 | |
| Sodium periodate | |
| [Na+].[O-][I](=O)(=O)=O |
| INaO4 | |
| JQWHASGSAFIOCM-UHFFFAOYSA-M | |
| sodium periodate |
Safety and Handling
Recommended Storage : Store at 5 to 30°C