missing translation for 'onlineSavingsMsg'
Learn More

Sodium metaperiodate, 99.0%, for analysis EMSURE™ ACS, Reag. Ph Eur, MilliporeSigma™

Catalog No. p-7162460
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1065970250 250 g
361041793 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1065970250 Supplier Technidata Supplier No. 1.06597.0250
Only null left

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym Sodium periodate
IUPAC Name sodium periodate
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Melting Point 270°C (decomposes)
pH 5.2 (50 g/L, H2O, 20°C)
Quantity 250 g
Assay Percent Range Assay (iodometric): ≥ 99.0%; Assay (out of dried substance): 99.8 to 100.3%
Solubility Information 91 g/L
Packaging Plastic Bottle
Density 3.87 g/cm3 (at 20°C); Bulk Density: 2900 kg/m3
Recommended Storage Store at 5 to 30°C