missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Metabisulfite, Granular, FCC, 90-100.5%, Spectrum™ Chemical
SO181, 7681-57-4, Na2S2O5
$376.03 - $5207.47
Chemical Identifiers
| CAS | 7681-57-4 |
|---|---|
| Molecular Formula | Na2O5S2 |
| Molecular Weight (g/mol) | 190.09 |
| MDL Number | MFCD00167602 |
| InChI Key | LDTLADDKFLAYJA-UHFFFAOYSA-L |
| IUPAC Name | disodium (sulfinatooxy)sulfinate |
| SMILES | [Na+].[Na+].[O-]S(=O)OS([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606648
|
Spectrum Chemical Mfg Cor
SO181500GM |
500 g | Poly Bottle |
Each for $376.03
|
|
||||
|
18606649
|
Spectrum Chemical Mfg Cor
SO18125KG |
2.5 kg | Poly Bottle |
Each for $1,043.20
|
|
||||
|
18606650
|
Spectrum Chemical Mfg Cor
SO18112KG |
12 kg | Poly Pail |
Each for $3,115.40
|
|
||||
|
18606651
|
Spectrum Chemical Mfg Cor
SO18150KGBL |
50 kg | Fiber Drum |
Each for $5,207.47
|
|
||||
Description
Spectrum™ Chemical Sodium Metabisulfite, Granular, FCC is used as a preservative and antioxidant in food and is also known as E223. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 7681-57-4 | |
| 190.09 | |
| LDTLADDKFLAYJA-UHFFFAOYSA-L | |
| [Na+].[Na+].[O-]S(=O)OS([O-])=O |
| Na2O5S2 | |
| MFCD00167602 | |
| disodium (sulfinatooxy)sulfinate |
Specifications
| 7681-57-4 | |
| 500 g | |
| MFCD00167602 | |
| [Na+].[Na+].[O-]S(=O)OS([O-])=O | |
| 190.09 | |
| FCC |
| 1 | |
| Na2O5S2 | |
| LDTLADDKFLAYJA-UHFFFAOYSA-L | |
| disodium (sulfinatooxy)sulfinate | |
| 90 to 100.5% | |
| Poly Bottle |