missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Metabisulfite, Granular, FCC, 90-100.5%, Spectrum™ Chemical
SO181, 7681-57-4, Na2S2O5
Supplier: Spectrum Chemical Mfg Cor SO18112KG
Description
Spectrum™ Chemical Sodium Metabisulfite, Granular, FCC is used as a preservative and antioxidant in food and is also known as E223. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 7681-57-4 | |
| 12 kg | |
| MFCD00167602 | |
| [Na+].[Na+].[O-]S(=O)OS([O-])=O | |
| 190.09 | |
| FCC |
| 1 | |
| Na2O5S2 | |
| LDTLADDKFLAYJA-UHFFFAOYSA-L | |
| disodium (sulfinatooxy)sulfinate | |
| 90 to 100.5% | |
| Poly Pail |
Chemical Identifiers
| 7681-57-4 | |
| 190.09 | |
| LDTLADDKFLAYJA-UHFFFAOYSA-L | |
| [Na+].[Na+].[O-]S(=O)OS([O-])=O |
| Na2O5S2 | |
| MFCD00167602 | |
| disodium (sulfinatooxy)sulfinate |