missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Lauryl Sulfate, Reagent, 97%, Spectrum™ Chemical
S1333, 151-21-3, C12H25O4SNa
$688.29 - $15,814.14
Chemical Identifiers
| CAS | 151-21-3 |
|---|---|
| Molecular Formula | C12H25NaO4S |
| Molecular Weight (g/mol) | 288.38 |
| MDL Number | MFCD00036175 |
| InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
| IUPAC Name | sodium dodecyl sulfate |
| SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18606623
|
N/A | 100 g | Amber Glass Bottle |
Each for $688.29
|
|
||||
|
18606624
|
N/A | 500 g | Amber Glass Bottle |
Each for $1,598.43
|
|
||||
|
18606625
|
N/A | 1 kg | Amber Glass Bottle |
Each for $3,005.25
|
|
||||
|
18606626
|
N/A | 12 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Sodium Lauryl Sulfate, Reagent , is an organic compound, and anionic surfactant derived from coconut or palm oils, for use as a detergent. As an Organosulfate, this salt has the amphiphilic properties needed in a detergent. It's uses vary from shark repellent, fruit fly killer and biocide. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Chemical Identifiers
| 151-21-3 | |
| 288.38 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| C12H25NaO4S | |
| MFCD00036175 | |
| sodium dodecyl sulfate |
Specifications
| 151-21-3 | |
| C12H25NaO4S | |
| MFCD00036175 | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 288.38 | |
| Reagent |
| 1 | |
| 0.05 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| sodium dodecyl sulfate | |
| 97% | |
| Sodium Lauryl Sulfate |