Learn More
Sodium Lauryl Sulfate, Reagent, 97%, Spectrum™ Chemical
S1333, 151-21-3, C12H25O4SNa
Supplier: Spectrum Chemical Mfg Cor S1333500GM
| Quantity | 500 g |
|---|---|
| Packaging | Amber Glass Bottle |
Description
Spectrum™ Chemical Sodium Lauryl Sulfate, Reagent , is an organic compound, and anionic surfactant derived from coconut or palm oils, for use as a detergent. As an Organosulfate, this salt has the amphiphilic properties needed in a detergent. It's uses vary from shark repellent, fruit fly killer and biocide. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Chemical Identifiers
| 151-21-3 | |
| 288.38 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| C12H25NaO4S | |
| MFCD00036175 | |
| sodium dodecyl sulfate |
Specifications
| Sodium Lauryl Sulfate | |
| 1 | |
| Amber Glass Bottle | |
| 0.05 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| sodium dodecyl sulfate | |
| 97% |
| 151-21-3 | |
| 500 g | |
| C12H25NaO4S | |
| MFCD00036175 | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 288.38 | |
| Reagent |