missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Lauryl Sulfate, NF, EP, BP, JP, Spectrum™ Chemical
SO180, 151-21-3, C12H25O4SNa
$7,466.25 - $18,987.04
Chemical Identifiers
| CAS | 151-21-3 |
|---|---|
| Molecular Formula | C12H25NaO4S |
| Molecular Weight (g/mol) | 288.38 |
| MDL Number | MFCD00036175 |
| InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
| IUPAC Name | sodium dodecyl sulfate |
| SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18606620
|
N/A | 12 kg | Fiber Drum |
Each for $7,466.25
|
|
||||
|
18606621
|
N/A | 45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Sodium Lauryl Sulfate, NF, EP, BP, JP is used as an emulsifying agent, modified-release agent, penetration enhancer, solubilizing agent, tablet and capsule lubricant. All Spectrum Chemical NF, EP, BP, and JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Chemical Identifiers
| 151-21-3 | |
| 288.38 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
| C12H25NaO4S | |
| MFCD00036175 | |
| sodium dodecyl sulfate |
Specifications
| 151-21-3 | |
| C12H25NaO4S | |
| MFCD00036175 | |
| [Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
| 288.38 | |
| Multi-Compendial |
| 1 | |
| 0.05 | |
| DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
| sodium dodecyl sulfate | |
| 85% | |
| Sodium Lauryl Sulfate |