missing translation for 'onlineSavingsMsg'
Learn More

Sodium Lauryl Sulfate, NF, EP, BP, JP, Spectrum™ Chemical

SO180, 151-21-3, C12H25O4SNa

Supplier:  Spectrum Chemical Mfg Cor SO18045KGBL

Quantity 45 kg
Packaging Fiber Drum

 View more versions of this product

Catalog No. 18606621

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Spectrum™ Chemical Sodium Lauryl Sulfate, NF, EP, BP, JP is used as an emulsifying agent, modified-release agent, penetration enhancer, solubilizing agent, tablet and capsule lubricant. All Spectrum Chemical NF, EP, BP, and JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

151-21-3
288.38
DBMJMQXJHONAFJ-UHFFFAOYSA-M
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
C12H25NaO4S
MFCD00036175
sodium dodecyl sulfate

Specifications

Sodium Lauryl Sulfate
1
Fiber Drum
0.05
DBMJMQXJHONAFJ-UHFFFAOYSA-M
sodium dodecyl sulfate
85%
151-21-3
45 kg
C12H25NaO4S
MFCD00036175
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
288.38
Multi-Compendial
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions