missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Iodide, USP, 99-101.5%, Spectrum™ Chemical
SO175, 7681-82-5, NaI
$193.70 - $42271.63
Chemical Identifiers
| CAS | 7681-82-5 |
|---|---|
| Molecular Formula | INa |
| Molecular Weight (g/mol) | 149.89 |
| InChI Key | FVAUCKIRQBBSSJ-UHFFFAOYSA-M |
| IUPAC Name | sodium iodide |
| SMILES | [Na+].[I-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606601
|
Spectrum Chemical Mfg Cor
SO1755GM |
5 g | Amber Glass Bottle |
Each for $193.70
|
|
||||
|
18606602
|
Spectrum Chemical Mfg Cor
SO17525GM |
25 g | Amber Glass Bottle |
Each for $307.00
|
|
||||
|
18606596
|
Spectrum Chemical Mfg Cor
SO175125GM |
125 g | Amber Glass Bottle |
Each for $569.90
|
|
||||
|
18606597
|
Spectrum Chemical Mfg Cor
SO175500GM |
500 g | Amber Glass Bottle |
Each for $1,715.03
|
|
||||
|
18606598
|
Spectrum Chemical Mfg Cor
SO17525KG |
2.5 kg | Amber Glass Bottle |
Each for $4,914.10
|
|
||||
|
18606599
|
Spectrum Chemical Mfg Cor
SO17512KG |
12 kg | Poly Pail |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18606600
|
Spectrum Chemical Mfg Cor
SO17545KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||