missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodide, USP, 99-101.5%, Spectrum™ Chemical

Catalog No. 18606601
Change view
Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606601 5 g Amber Glass Bottle
18606602 25 g Amber Glass Bottle
18606596 125 g Amber Glass Bottle
18606597 500 g Amber Glass Bottle
18606598 2.5 kg Amber Glass Bottle
18606599 12 kg Poly Pail
18606600 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18606601 Supplier Spectrum Chemical Mfg Cor Supplier No. SO1755GM

SO175, 7681-82-5, NaI

Spectrum™ Chemical Sodium Iodide, USP is a crystalline salt that is white and is used as a food supplement to counter and prevent iodine deficiencies. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.89
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
IUPAC Name sodium iodide
SMILES [Na+].[I-]

Specifications

CAS Max % 1
Quantity 5 g
Percent Purity 99 to 101.5%
Grade USP
Packaging Amber Glass Bottle