missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodide, USP, 99-101.5%, Spectrum™ Chemical

SO175, 7681-82-5, NaI

Supplier:  Spectrum Chemical Mfg Cor SO175500GM

 View more versions of this product

Catalog No. 18606597

Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Sodium Iodide, USP is a crystalline salt that is white and is used as a food supplement to counter and prevent iodine deficiencies. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

7681-82-5
500 g
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
sodium iodide
99 to 101.5%
Amber Glass Bottle
1
INa
[Na+].[I-]
149.89
USP

Chemical Identifiers

7681-82-5
149.89
sodium iodide
INa
FVAUCKIRQBBSSJ-UHFFFAOYSA-M
[Na+].[I-]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions