missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodate, Powder, 99.5%, Spectrum™ Chemical

$267.22 - $31231.88

Chemical Identifiers

CAS 7681-55-2
View More Specs

Products 7
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613682
View Documents
Spectrum Chemical Mfg Cor
S132525GM
25 g Amber Glass Bottle
Each for $267.22
Only null left
 
18613679
View Documents
Spectrum Chemical Mfg Cor
S1325100GM
100 g Amber Glass Bottle
Each for $505.59
Only null left
 
18613680
View Documents
Spectrum Chemical Mfg Cor
S1325125GM
125 g Amber Glass Bottle
Each for $601.44
Only null left
 
18613683
View Documents
Spectrum Chemical Mfg Cor
S1325500GM
500 g Amber Glass Bottle
Each for $1,370.81
Only null left
 
18613685
View Documents
Spectrum Chemical Mfg Cor
S132525KG
2.5 kg Amber Glass Bottle
Each for $3,830.16
Only null left
 
18613681
View Documents
Spectrum Chemical Mfg Cor
S132512KG
12 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18613684
View Documents
Spectrum Chemical Mfg Cor
S132550KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7681-55-2

Specifications

7681-55-2
25 g
Reagent
Amber Glass Bottle
1
99.5%
0.003
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.