accessibility menu, dialog, popup

UserName

NaOIOO

CAS RN 7681-55-2

IUPAC Name: sodium iodate
Synonyms: sodium iodate
Molecular Weight (g/mol): 197.89
Molecular Formula: INaO3
InChi Key: WTCBONOLBHEDIL-UHFFFAOYSA-M
SMILES: [Na+].[O-][I](=O)=O
Percent Purity 
  • (3)
  • (1)
  • (2)
  • (2)
  • (7)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
Grade 
  • (3)
  • (2)
  • (7)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Physical Form 
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (7)
  • (7)
  • (3)
  • (1)
  • (2)
  • (2)
  • (7)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (7)
  • (5)
  • (3)
  • (2)
  • (5)