missing translation for 'onlineSavingsMsg'
Learn More

Sodium Iodate, Powder, 99.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S1325100GM

 View more versions of this product

Catalog No. 18613679


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7681-55-2
100 g
Reagent
Amber Glass Bottle
1
99.5%
0.003

Chemical Identifiers

7681-55-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.