missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite Solution, 2.5% (w/w) NaOCl, Ricca Chemical

Supplier:  Ricca Chemical Company 7495132

 View more versions of this product

Catalog No. 7495-132


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Hypochlorite Solution
7732-18-5,7681-52-9
99.0,2.23
1 L
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
[O-]Cl.[Na+]
74.439
CHEBI:32146
Laboratory
Natural Poly Bottle
Colorless to Green-Yellow
97.0,2.15
Liquid
ClNaO
SUKJFIGYRHOWBL-UHFFFAOYSA-N
sodium;hypochlorite
23665760
2.5% W/W
2.5% (w/w) NaOCl

Chemical Identifiers

7732-18-5
74.439
sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
CHEBI:32146
[O-]Cl.[Na+]
ClNaO
SUKJFIGYRHOWBL-UHFFFAOYSA-N
23665760
sodium;hypochlorite
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 6 months

SDS
Documents

Documents

Promotions

Promotions