missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hypochlorite Solution, 2.5% (w/w) NaOCl, Ricca Chemical
Specifications
Specifications
| Color | Colorless to Green-Yellow |
| CAS Min % | 97.0, 2.15 |
| CAS Max % | 99.0, 2.23 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Percent Purity | 2.5% W/W |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 2.5% (w/w) NaOCl |
| Packaging | Natural Poly Bottle |
| Chemical Name or Material | Sodium Hypochlorite Solution |
Safety and Handling
| ShelfLife | 6 months |