missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite, 5% Available Chlorine, Solution, Reagent, Spectrum™ Chemical

Catalog No. 18606586
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606586 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606586 Supplier Spectrum Chemical Mfg Cor Supplier No. S1315500MLPL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1315, 7681-52-9, NaOCl

Spectrum™ Chemical Sodium Hypochlorite, 5 Percent Available Chlorine, Solution, Reagent is made from passing chlorine gas through cold and dilute sodium hydroxide solution. The primary use of Sodium Hypochlorite in labs and manufacturing is in surface purification, textile bleaching, odor removal in water treatment facilities, and to detoxify cyanide baths in the metal processing industry.

Chemical Identifiers

CAS 7681-52-9 , 7732-18-5 , 1310-73-2
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.44
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
IUPAC Name sodium hypochlorite
SMILES [Na+].[O-]Cl

Specifications

CAS Max % 0.07, 0.92, 0.01
Physical Form Solution
Quantity 500 mL
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.07
Chemical Name or Material Sodium Hypochlorite, 5 Percent Available Chlorine, Solution