missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite, 5% Available Chlorine, Reagents

Sodium Hypochlorite, 5% Available Chlorine, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

$58.20 - $158.43

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Bleach
IUPAC Name water
SMILES O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-337-009
View Documents
Labchem Inc
C23190001A
1 L
Each for $58.20
Only null left
 
01-337-008
View Documents
Labchem Inc
C23190004A
4 L
Each for $158.43
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
18.02
Bleach
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water

Specifications

40°C
Colorless to Greenish-Yellow
Liquid
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
5 to 6%
Poly Bottle
Sodium Hypochlorite, 5% Available Chlorine
0°C
7732-18-5,7681-52-9
1 L
Bleach
O
18.02
Technical
1.08
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'