missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hypochlorite, 5% Available Chlorine, Reagents

Catalog No. 01337009
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337009 1 L
01337008 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337009 Supplier Labchem Inc Supplier No. C23190001A
Only null left

Sodium Hypochlorite, 5% Available Chlorine, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7681-52-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Bleach
IUPAC Name water
SMILES O

Specifications

Boiling Point 40°C
Melting Point 0°C
Color Colorless to Greenish-Yellow
Physical Form Liquid
Quantity 1 L
Percent Purity 5 to 6%
Grade Technical
Packaging Poly Bottle
Specific Gravity 1.08
Chemical Name or Material Sodium Hypochlorite, 5% Available Chlorine