missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution 50%, for analysis EMSURE™, MilliporeSigma™

Catalog No. 361041237
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361041237 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361041237 Supplier Technidata Supplier No. 1.58793.1000
Only null left

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

Boiling Point 143°C
Density 1.53g/cm3 (20°C)
Packaging Plastic Bottle
Quantity 1 L
Chemical Name or Material Sodium hydroxide solution
Recommended Storage Store at 15° to 25°C