missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide solution, 5.0M, Solstice
Description
Solstice's high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Linear Formula | NaOH |
| Concentration | 5.0M |
| UN Number | UN1824 |
| Density | 1.182g/mL at 25°C |
| Quantity | 6 x 1 L |
| Formula Weight | 40g/mol |
| Grade | Volumetric |
| Packaging | Plastic Bottle |
| Vapor Pressure | 3 mmHg (37°C) |
| Chemical Name or Material | Sodium hydroxide solution |
Safety and Handling
| Hazard Statement | P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 |