missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Solution, 40%, Fisher Chemical™

Catalog No. p-2519655
Change view
Click to view available options
Quantity:
4 L
10 L
Packaging:
Poly Bottle
PolyPac™
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SS4114 4 L Poly Bottle
SS41110 10 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SS4114 Supplier Thermo Fisher Scientific Supplier No. SS411 4
Only null left

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

pH 12.0
Boiling Point 141.7°C
Melting Point 5°C
Quantity 4 L
Percent Purity 40 to 42%
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure 3mmHg at 37°C
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide Solution, 40% (w/w)
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Causes severe burns by all exposure routes. Wear personal protective equipment. Ensure adequate ventilation. Do not breathe vapors or spray mist. Do not get in eyes, on skin, or on clothing. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:3
Flammability:0
Instability:1
DOTInformation DOT Class 8, : Corrosive